Learn More
Cell Biolabs Inc CytoSelect™ 96-Well Phagocytosis Assay Kit (Red Blood Cell, Colorimetric), 96 assays
Supplier:  Cell Biolabs Inc CBA220
Encompass_Preferred
Erythrocytes (RBCs) are commonly used in phagocytosis assays. For FcR mediated phagocytosis, RBCs are opsonized with serum or IgG before they are added to phagocytes. After removal of non-phagocytic erythrocytes, engulfed RBCs per phagocyte cell are manually counted (expressed as phagocytosis index or engulfed RBCs per phagocyte). This manual counting method is difficult when testing a large number of samples. CytoSelect™ 96-well Phagocytosis Assay does not involve subjective manual counting of RBCs. Instead cells are lysed and detected by the proprietary erythrocyte substrate in a microtiter plate reader. This format provides a quantitative, high-throughput method to accurately measure phagocytosis. The CytoSelect™ 96-well Phagocytosis Assay provides a robust system for screening TLR ligands, phagocytosis activators or inhibitors. Each kit provides sufficient quantities to perform 96, 48, 24 tests in a 96, 48, or 24-well plate, respectively. RBCs not included.
Catalog No. 50-672-671
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.