Learn More
Abcam Phalloidin FITC Reagent, Fluorescence Microscopy, Liquid, 300 Tests
Description
- Applications: Fluorescence Microscopy, ICC/IF
- Form: Liquid
- Composition: 99.9% Dimethylsulfoxide, 0.1% Phalloidin-Fluorescein Conjugate
- Usage: Suitable for staining of F-actin filaments in tissue sections and cell cultures
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | F-actin |
| Buffer | 99.9% dimethylsulfoxide, 0.1% phalloidin-fluorescein conjugate |
| Form | Liquid |
| Kit Contents | Phalloidin-Fluorescein Conjugate (0.1%), Dimethylsulfoxide (99.9%) |
| Label or Dye | FITC |
| No. of Reactions | 300 Tests |
| Substrate Properties | Chemical Substrate |
| Substrate | F-actin |
| Detection Method | Fluorescence |
| Formulation | 99.9% Dimethylsulfoxide, 0.1% Phalloidin-Fluorescein Conjugate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.