Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Phalloidin-iFluor 350 Reagent, Liquid, 300 Tests

Catalog No. 502582278
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2278 300 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2278 Supplier Abcam Supplier No. AB176751300TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent probe for the selective staining of F-actin in fluorescence microscopy and ICC/IF applications.

  • Applications: Fluorescence Microscopy, ICC/IF
  • Form: Liquid
  • Storage buffer: 99% Dimethylsulfoxide and 1% Fluorescent-Phalloidin Conjugate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate F-actin
Substrate Properties Peptide-based Substrate
Target Molecule F-actin
Kit Contents Phalloidin-iFluor 350 Conjugate (1% in 99% Dimethylsulfoxide)
No. of Reactions 300 Tests
Label or Dye iFluor 350
For Use With (Equipment) Fluorescence Microscope
Form Liquid
Buffer 99% dimethylsulfoxide, 1% fluorescent-phalloidin conjugate
Detection Method Fluorescence
Formulation 99% Dimethylsulfoxide, 1% Fluorescent-Phalloidin Conjugate
Quantity 300 Tests
For Use With (Application) Fluorescence Microscopy, ICC/IF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.