Learn More
Aat Bioquest Phalloidin-iFluor 488 Conjugate (300 Tests)
SDP
Supplier:  Aat Bioquest 23115
Encompass_Preferred
This green fluorescent phalloidin conjugate equivalent to Alexa Fluor 488-labeled phalloidin selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates. This catalog is for 300 Tests.
Catalog No. 50-225-0144
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.