Learn More
Cayman Chemical Isobavachalcon 5mg
SDP
Supplier:  Cayman Chemical 198735 MG
Encompass_Preferred
A selective mGluR1 PAM (EC50s 0 39 and 0 36 M for human and rat receptors respectively) with no activity as a mGlu4 PAM (EC50 10 M) potentiates a response to glutamate in wild-type cell lines stably expressing mGlu1 and partially restores the reduction in glutamate-mediated calcium signaling in a mutant cell model of schizophrenia
Catalog No. 50-298-2807
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.