Learn More
Strem, An Ascensus Company CAS 849950-56-7. 500mg. (R)-(-)-412-Bis(diphenylphosphino)[2.2]paracyclophane(15-cyclooctadiene)rhodium(I) tetrafluoroborate min. 97%. MFCD16294983
SDP
Supplier:  Strem, An Ascensus Company 450217500MG
Encompass_Preferred
CAS 849950-56-7. 500mg. (R)-(-)-412-Bis(diphenylphosphino)[2.2]paracyclophane(15-cyclooctadiene)rhodium(I) tetrafluoroborate min. 97%. MFCD16294983. Molecular Weight 874.54. Molecular Formula C48H46BF4P2Rh. Color/form yellow-orange solid Strem 45-0217. http//www.strem.com/catalog/v/45-0217/
Catalog No. 50-211-0841
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.