Learn More
Foamtec PharmaMOP™ Cleanroom Mop Heads, Sterile
Catalog No. 19304821 Shop All Foamtec International Products
Change view
Click to view available options
Material:
Microfiber
Polyester
Product Type:
EZ Glide Gamma Irradiated Mop Head
EZ Glide Mop Head
Gamma Irradiated Mop Head
High Fluid Capacity Cleanroom Mop Head
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
19-304-821 Polyester EZ Glide Gamma Irradiated Mop Head
19-304-820 Polyester Gamma Irradiated Mop Head
19-304-822 Polyester High Fluid Capacity Cleanroom Mop Head
19-304-823 Polyester EZ Glide Mop Head
19-304-825 Microfiber EZ Glide Mop Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Catalog No. 19-304-821 missing translation for 'mfr' Foamtec missing translation for 'supplierNo' FS853ENST
For cleaning and disinfection of rough surfaces and high soil areas
Enables the quick, efficient cleaning of surfaces stained with disinfectant residues
  • Gamma irradiated
  • EzGLIDE™ polyester mop head
  • 0.125 in. foam core
Certifications
ISO 9001:2000
Specifications
Product Type EZ Glide Gamma Irradiated Mop Head
Sterility Sterile
For Use With (Application) For cleaning and disinfection of rough surfaces and high soil areas
Material Polyester