Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec PharmaMOP™ Cleanroom Mop Heads, Sterile

Catalog No. 19304825 Shop All Foamtec International Products
Change view
Click to view available options
Material:
Microfiber
Polyester
Product Type:
EZ Glide Gamma Irradiated Mop Head
EZ Glide Mop Head
Gamma Irradiated Mop Head
High Fluid Capacity Cleanroom Mop Head
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type
19-304-825 Microfiber EZ Glide Mop Head
19-304-820 Polyester Gamma Irradiated Mop Head
19-304-822 Polyester High Fluid Capacity Cleanroom Mop Head
19-304-823 Polyester EZ Glide Mop Head
19-304-821 Polyester EZ Glide Gamma Irradiated Mop Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-304-825 Supplier Foamtec Supplier No. FS858ENST

For cleaning and disinfection of rough surfaces and high soil areas

Enables the quick, efficient cleaning of surfaces stained with disinfectant residues

  • Gamma irradiated
  • EzGLIDE™ polyester mop head
  • 0.125 in. foam core

Certifications

ISO 9001:2000

Specifications

Product Type EZ Glide Mop Head
Sterility Sterile
Size Large (1.45 lb.)
For Use With (Application) For cleaning and disinfection of rough surfaces and high soil areas, steam sterilization cycle
Certifications/Compliance ISO 9001:2000
Material Microfiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.