Learn More
CPC Scientific H-Pro-His-Cys-Lys-Arg-Met-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific AOXP001B
Encompass
A synthetic oligopeptide containing cysteine and histidine, PHCKRM, shows ability to scavenge oxygen from superoxide (O2-) and hydroxyl radical (.OH). The antioxidant effect of the peptide may mitigate cancers that involve hydroxyl radical damage.SEQUENCE: H-Pro-His-Cys-Lys-Arg-Met-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: PHCKRMMOLECULAR FORMULA: C31H54N12O7S2MOLECULAR WEIGHT: 770.98STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [159147-88-3]SYNONYMS: RESEARCH AREA: CancerREFERENCES:J.Ueda et al., Biochem. Mol. Biol. Int. , 33, 1041 (1994)SKU(s): AOXP-001A, AOXP-001B
Catalog No. 50-210-8771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.