Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenanthrene Solution, 100μg/mL in Dichloromethane, Ultra Scientific

Catalog No. USP7501
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-P7501 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-P7501 Supplier Agilent Technologies Supplier No. P7501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-01-8 , 75-09-2
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

CAS Max % 99.99, 0.01
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Phenanthrene Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.