CAS RN 85-01-8
Phenanthrene, 97%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene, tech. 90%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene, 98%
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene 97.0+%, TCI America™
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Agilent Technologies Phenanthrene Solution, 100μg/mL in Dichloromethane, Ultra Scientific
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Sigma Aldrich Phenanthrene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Phenanthrene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Phenanthrene, MP Biomedicals
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene 95.0+%, TCI America™
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N Synonym: phenanthren,phenanthrin,phenanthracene,ravatite,phenantrin,phenanthren german,phenanthrene, pure,unii-448j8e5bst,ccris 1233,chembl46730 PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1
Phenanthrene, SPEX CertiPrep™
CAS: 85-01-8 Molecular Formula: C14H10 Molecular Weight (g/mol): 178.23 MDL Number: MFCD00001168 InChI Key: YNPNZTXNASCQKK-UHFFFAOYSA-N PubChem CID: 995 ChEBI: CHEBI:28851 IUPAC Name: phenanthrene SMILES: C1=CC=C2C(C=CC3=CC=CC=C23)=C1