Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenanthrene 95.0+%, TCI America™
SDP

Catalog No. P131025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P131025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P131025G Supplier TCI America Supplier No. P131025G

Chemical Identifiers

CAS 85-01-8
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001168
InChI Key YNPNZTXNASCQKK-UHFFFAOYSA-N
Synonym phenanthren, phenanthrin, phenanthracene, ravatite, phenantrin, phenanthren german, phenanthrene, pure, unii-448j8e5bst, ccris 1233, chembl46730
PubChem CID 995
ChEBI CHEBI:28851
IUPAC Name phenanthrene
SMILES C1=CC=C2C(C=CC3=CC=CC=C23)=C1

Specifications

Melting Point 100°C
Color White-Yellow
Boiling Point 340°C
Quantity 25 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Phenanthrene
EINECSNumber (4)-0635
RTECSNumber SF7175000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.