Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazine ethosulfate, ∽95%, MP Biomedicals™

Catalog No. ICN10095601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10095601 1 g
ICN10095605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10095601 Supplier MP Biomedicals, Inc Supplier No. 0210095601
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10510-77-7
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
MDL Number MFCD00050286
InChI Key VDJKJPMLWJWQIH-UHFFFAOYSA-M
Synonym phenazine ethosulfate, 5-ethylphenazine, n-ethylphenazonium ethosulfate, 5-ethylphenazinium ethyl sulphate, 5-ethylphenazin-5-ium ethyl sulfate, 5-ethylphenazinium ethylsulfate, phenazinium, 5-ethyl-, ethyl sulfate, 5-ethylphenazine, ethyl sulfate salt, ccris 5619, n-ethyldibenzopyrazine ethyl sulfate salt
PubChem CID 82688
IUPAC Name 5-ethylphenazin-5-ium;ethyl sulfate
SMILES CC[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.CCOS(=O)(=O)[O-]

Specifications

Color Orange-Yellow
Assay Percent Range ∽ 95%
Quantity 1 g
Formula Weight 334.4
Physical Form Powder
Chemical Name or Material Phenazine ethosulfate
Recommended Storage Store at -0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.