Learn More
Apexbio Technology LLC Phenazine methosulfate 299-11-6 100mg
SDP
Supplier:  Apexbio Technology LLC C7048100
Encompass_Preferred
Phenazine methosulfate (299-11-6) is a redox mediator that facilitates electron transfer reactions resulting in the generation of free radicals and reactive oxygen species (ROS) Phenazine methosulfate exerts its biological activity primarily through the transfer of electrons from NADPH leading to oxidative DNA damage single-strand DNA breakage and the induction of apoptotic pathways In in vitro studies phenazine methosulfate is commonly used to assess cellular viability quantify oxidative stress and DNA damage responses and investigate mechanisms of apoptosis Reported cytotoxic activity varies among cell types with typical IC50 values ranging from approximately 5 M to 50 M depending on experimental conditions Based on these pharmacological properties phenazine methosulfate holds research potential in anti-tumor assays DNA damage studies oxidative stress modeling and pest control applications
Catalog No. 50-000-90075
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.