Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phencyclidine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175340
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175340 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175340 Supplier Invitrogen™ Supplier No. PA175340
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 1/16K. Sensitivity: 10 ng/mL PCP produces 89% inhibition in a competitive ELISA employing Phencyclidine (piperidyl,4) polyclonal antibody.

Phencyclidine is an illegal, hallucinogenic drug that was initially used as an anesthetic agent in the 1950s and early 1960s, but was then withdrawn in 1965 because of dissociative hallucinogenic effects that were often disturbing and sometimes severe and prolonged. The out-of-body intense psychological and behavioral effects of low doses of phencyclidine led to its abuse. In the late 1960s and 1970s, phencyclidine (angel dust) became a widely used hallucinogenic drug. The effects were often extreme, marked by acute psychosis and aggressive and violent behaviors, and overdoses led to many emergency room visits and deaths from status epilepticus, hyperthermia, rhabdomyolysis and subsequent renal, respiratory and hepatic failure. [National Center for Biotechnology Information. PubChem Compound Database; CID=6468].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phencyclidine
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias C17H25N; PCP
Host Species Sheep
Immunogen PCP(piperidyl,4)-BSA.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.