Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T027025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T027025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T027025G Supplier TCI America Supplier No. T027025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4455-09-8
Molecular Formula C15H16O3S
Molecular Weight (g/mol) 276.35
MDL Number MFCD00059322
InChI Key CVPPUZPZPFOFPK-UHFFFAOYSA-N
Synonym 2-Phenylethyl p-Toluenesulfonate, p-Toluenesulfonic Acid Phenethyl Ester, p-Toluenesulfonic Acid 2-Phenylethyl Ester
PubChem CID 20522
IUPAC Name 2-phenylethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC2=CC=CC=C2

Specifications

Melting Point 38°C
Color White
Quantity 25 g
Formula Weight 276.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenethyl p-Toluenesulfonate
EINECSNumber (3)-1934
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.