Learn More
Selleck Chemical LLC Phenformin HCl S2542-1g
SDP
Supplier:  Selleck Chemical LLC S25421G
Encompass_Preferred
Phenformin HCl is a hydrochloride salt of phenformin that is an anti-diabetic drug from the biguanide class It activates AMPK increasing activity and phosphorylation
Catalog No. 50-311-1668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.