Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenobarbital Monoclonal Antibody (PB9-30.17.4.16), QED Bioscience™
SDP

Catalog No. 502277692
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7692 1 mg
50-227-7693 100 μg
50-227-7694 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7692 Supplier Leinco Technologies Inc Supplier No. 164011000UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phenobarbital Monoclonal antibody specifically detects Phenobarbital in Chemical samples. It is validated for ELISA

Specifications

Antigen Phenobarbital
Applications ELISA
Classification Monoclonal
Clone PB9-30.17.4.16
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C12H12N2O3; Luminal; Phenobarbitol; Phenobarbitone; Phenylethylbarbiturate
Host Species Mouse
Immunogen Phenobarbital-KLH.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.