Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol Stock Standard, 5.00 ± 0.05% (w/v), Ricca Chemical
SDP

Catalog No. 57381 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57381 4 L
5738-16 500 mL
5738-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 57381 Supplier Ricca Chemical Company Supplier No. 57381
Add to Cart
Add to Cart

AOAC/APHA for Selenium Testing

Specifications

Chemical Name or Material Phenol Stock Standard
Packaging Amber Glass Bottle
CAS 108-95-2, 7732-18-5
Concentration or Composition Notes 5.00 ┬▒ 0.05% (w/v)
CAS Min % 93.0, 5.27
CAS Max % 97.0, 5.49
Physical Form Liquid
Molecular Formula C6H6O
MDL Number MFCD00002143
Color Colorless
Melting Point Approximately 0°C
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
Type Standard
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
Solvent or Matrix Water
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
Quantity 4 L
PubChem CID 996
ChEBI CHEBI:15882
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.