Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, 99%, extra pure

Catalog No. AC149340025
Change view
Click to view available options
Quantity:
50 g
1 kg
2.5 kg
5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149340025 2.5 kg Glass bottle
AC149340500 50 g Glass bottle
AC149340010 1 kg Glass bottle
AC149340051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC149340025 Supplier Thermo Scientific Chemicals Supplier No. 149340025
Add to Cart

CAS: 108-95-2 | C6H6O | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39°C to 42°C
Color Colorless
Density 1.0700g/mL
Boiling Point 182°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OH
Quantity 2.5 kg
Beilstein 06, 110
Fieser 01,828; 13,166; 14,88
Specific Gravity 1.07
Merck Index 15, 7354
Solubility Information Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides,soluble in alcohol,chloroform,ether,glycerol,,carbon disulfide and fixed oils,1g/12mL benzene,practically insoluble in petroleum ether
Viscosity 3.437 mPa.s (50°C)
Vapor Pressure 0.4mbar at 20°C
Formula Weight 94.11
Percent Purity 99%
Grade Extra Pure
Physical Form Crystals or Fused Mass
Chemical Name or Material Phenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection
IF SWALLOWED: rinse mouth. Do NOT induce vomiting
IF ON SKIN: Gently wash with plenty of soap and water
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Immediately call a POISON CENTER or doctor/physician
EINECSNumber 203-632-7
Recommended Storage Turns pink on exposure to air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.