Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Phenol Crystals
SCI_ED

Catalog No. S25462 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25462 100 g Lab Grade
S25462A 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25462

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym Carbolic Acid, Phenyl Hydroxide
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Assay Percent Range 99%
Linear Formula C6H5OH
Formula Weight 94.11
Packaging Lab Grade
Density 1.0576g/mL
Color Colorless
Quantity 100 g
Chemical Name or Material Phenol
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 6.1, II, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.