Learn More
Specifications
Specifications
| Melting Point | 42.8°C |
| Color | Colorless |
| Density | 1.0576g/cm³ |
| pH | 6 |
| Boiling Point | 182°C |
| Flash Point | 79.4°C |
| Packaging | Lined Steel Drum |
| Quantity | 200 L |
| UN Number | 2821 |
| Residue after Evaporation | 0.05% max. |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Combustible liquid. Toxic by inhalation. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. NFPA Health:4 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.