Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol Red, Free Acid (Certified ACS), Fisher Chemical

Catalog No. P7410 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P74-10 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. P74-10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pH indicator

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym 4,4'-(3H-2,1-Benzoxathiol-3-yildene)bisphenol S,S-Dioxide, 4, 4'-(3H-2, 1-Benzoxathiol-3-yildene)bisphenol S
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 300°C
Clarity Pass Test
Identification Pass Test
Packaging Glass Bottle
Visual Transition Interval Pass Test
Linear Formula C6H4SO2OC(C6H4-4-OH)2
Color Red
Quantity 10 g
Physical Form Solid
Formula Weight 354.39
Grade Certified ACS
Chemical Name or Material Phenol Red, Free Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.