Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Phenol Red Indicator, ACS Grade
SCI_ED

Catalog No. s25465 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25465 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25465
Only null left
Add to Cart
Add to Cart

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
Synonym Phenolsulfonephthalein Sodium
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Assay Percent Range 97 to 100%
Linear Formula C19H13NaO5S
Formula Weight 376.36
Packaging ACS Grade
Density 1g/mL
pH Color Changes pH 6.8-8.2 (yellow-red)
Quantity 10 g
Chemical Name or Material Phenol Red Indicator
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.