Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phenol Red sodium salt, ACS reagent

Catalog No. AC417250100
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417250100 10 g Glass bottle
AC417250050 5 g Glass bottle
AC417251000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC417250100 Supplier Thermo Scientific Chemicals Supplier No. 417250100
Only null left
Add to Cart

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285.0°C
Clarity passes test
Beilstein 19, 91
Merck Index 15, 7360
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 376.35
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 6.8 (yellow) to pH 8.2 (red)
Color White
Quantity 10 g
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Phenol Red sodium salt
Show More Show Less
missing translation for 'healthHazard1' Warning
missing translation for 'healthHazard2' GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.