Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ KIMBLE™ Phenolic Screw Cap with Liner
GSA_VASDP

Catalog No. 055699 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
15-415
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
05-569-9 15-415
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-569-9 Supplier DWK LIFE SCIENCES™ Supplier No. 7380015415
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Secure your samples during autoclaving with the KIMBLE™ Phenolic Screw Cap with Liner. Made from specially formulated phenolic resin, it withstands repeated autoclaving and is resistant to stress cracking, offering superior torque retention for reliable laboratory use.

Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving

  • Specially formulated phenolic resin to withstand the effects of repeated autoclaving
  • Resistant to stress cracking
  • Designed for superior torque retention
  • Styrene liner provides a secure seal
  • Suitable for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 15-415
Liner Material Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Description Fixed Screw Cap
Material Phenolic
Color Black
For Use With (Application) For general laboratory use
Quantity 1000 Pack
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.