Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
GSA_VASDP

Catalog No. p-8003225 Shop All DWK Life Sciences Products
Change view
Click to view available options
Liner Material:
Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Liner Material
05-569-9 Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-569-9 Supplier DWK Life Sciences Supplier No. 7380015415
Only null left
Add to Cart
Add to Cart

Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving

  • Styrene Liner
  • Screw Cap

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Size 15-415
Liner Material Rubber
Product Type Screw Cap
Autoclavable Autoclavable
Description Fixed Screw Cap
Material Phenolic
Color Black
For Use With (Application) For general laboratory use
Quantity 1000 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.