Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, 1% (w/v) in 50% (v/v) Isopropyl Alcohol, Ricca Chemical
SDP

Catalog No. R56250004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
60 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R56250004A 4 L Natural Poly Bottle
R562500060A 60 mL Natural Poly Bottle
5625-16 500 mL Natural Poly Bottle
5625-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R56250004A Supplier Ricca Chemical Company Supplier No. 56251
Add to Cart
Add to Cart

Neutralized

Specifications

Chemical Name or Material Phenolphthalein
CAS 7732-18-5, 67-63-0, 77-09-8
CAS Min % 49.0, 0.5
CAS Max % 51.0, 1.5
Percent Purity 1% W/V
Grade Indicator
Packaging Natural Poly Bottle
Color Colorless to Pink
Quantity 4 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.