Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phenolphthalein, 2% soln. in 95% ethanol

Catalog No. AAJ61785AE
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61785AE 100 mL
AAJ61785AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61785AE Supplier Thermo Scientific Chemicals Supplier No. J61785AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

pH indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

It is used as a laboratory reagent and pH indicator. Phenolphthalein is a common indicator to use in neutralization titrations. Phenolphthalein exerts laxative effects by stimulating the intestinal mucosa and constricting smooth muscles. It is used medicinally as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Color Colorless
UN Number UN1987
Concentration 2% soln. in 95% ethanol
Merck Index 14,7243
Solubility Information Miscible in water.
Formula Weight 318.33
Percent Purity 95% (ethanol)
Odor Alcohol-like
Quantity 100 mL
Flash Point 17°C (63°F)
Physical Form Liquid
Chemical Name or Material Phenolphthalein
Show More Show Less
Hazard Category H225-H336-H341-H350-H361-H371
Hazard Statement GHS H Statement
H225-H350-H341-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 201-004-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.