Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, ACS Grade, Sigma-Aldrich

Catalog No. 01259058 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-058 100 g
01-258-807 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-058 Supplier MilliporeSigma Supelco Supplier No. 105945100G
Add to Cart
Add to Cart

Phenolphthalein is an indicator for acid-base titration reactions. It changes from colorless to violet-red between pH 8. 3 to 9. 8, it is red in extremely acidic solutions, and colorless in extremely alkaline solutions.

It is present as a nonionic, lipophilic lactone in aqueous neutral solutions. It exists as a hydrophilic dianion in alkaline solutions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Weight (g/mol) 318.32 g/mol

Specifications

Grade ACS
Color White
pH 8 to 10
Quantity 100 g
Chemical Name or Material Phenolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.