Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Diphosphate Pentasodium Salt Hydrate 95.0+%, TCI America™
SDP

Catalog No. P22931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22931G 1 g
P22935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22931G Supplier TCI America Supplier No. P22931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2382-56-1
Molecular Formula C20H18Na5O11P2
Molecular Weight (g/mol) 611.249
InChI Key BETACOHSDKIQSV-UHFFFAOYSA-N
PubChem CID 131876444
IUPAC Name 2-[hydroxy-bis(4-phosphonooxyphenyl)methyl]benzoic acid;sodium
SMILES C1=CC=C(C(=C1)C(=O)O)C(C2=CC=C(C=C2)OP(=O)(O)O)(C3=CC=C(C=C3)OP(=O)(O)O)O.[Na].[Na].[Na].[Na].[Na]

Specifications

Color Red-White
Quantity 1 g
Formula Weight 606.21
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein Diphosphate Pentasodium Salt Hydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.