Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Phenolphthalein Indicator
SCI_ED

Catalog No. p-4416725 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Reagent Grade - Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25466 25 g Reagent Grade - Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25466

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym 3, 3-Bis(4-Hydroxylphenyl)-1(3H)-Isobenzofuranone
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Assay Percent Range 100% w/v
Linear Formula C20H14O4
Formula Weight 318.33
Grade Reagent
Packaging Reagent Grade - Powder
Density 1.299g/mL
pH Color Changes pH 8.0-10.0 (colorless-red)
Color White
Quantity 25 g
Chemical Name or Material Phenolphthalein Indicator
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.