Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Phenolphthalein in IPA Solution
SCI_ED

Catalog No. S25468 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
1%
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25468 500 mL -
S25467 100 mL 1%
S25467A 500 mL 1%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S25468
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Specifications

Chemical Name or Material Phenolphthalein in IPA Solution
CAS 77-09-8, 7732-18-5
Formula Weight Mixture
Percent Purity 0.5%
Density 0.85 to 0.95
Color Colorless to Pink
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.