Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Stock Solution, Fisher Chemical™
SDP

Catalog No. SP901025 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP9010-25 25 mL Natural Poly Bottle
SP9010-50 50 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP9010-25
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Phenolphthalein Stock Solution
CAS 7732-18-5, 64-17-5, 67-56-1, 67-63-0, 77-09-8
CAS Min % 52.0, 43.0, 2.0, 2.0, 0.01
CAS Max % 55.0, 44.0, 3.0, 3.0, 0.01
Grade Indicator
Packaging Natural Poly Bottle
Color Colorless
pH 8
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.