Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Phenolphthalein Test Paper for Ammonia Detection - Ammonia Leak Detector Strips for Refrigerators, Air Conditioners, HVAC Maintenance, 100 Strips
SCI_ED

Catalog No. S02927 Shop All Eisco Products
Change view
Click to view available options
Range:
3 to 5 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S02927 3 to 5 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02927 Supplier Eisco™ Supplier No. CH202408
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eisco Labs Phenolphthalein Test Paper for Ammonia Detection. Designed for professional and home use, these strips maintain the integrity of refrigerators, air conditioners, and HVAC systems. Each pack contains 100 high-quality test strips, providing a reliable solution for detecting ammonia leaks.

  • Quick Ammonia DetectionTest: Paper changes to pinkish color when detecting ammonia vapors, ideal for leak checking.
  • Versatile Applications: Useful for maintenance checks in refrigerators, air conditioners, and industrial systems.
  • Sensitive and Accurate: Detects ammonia in solution from 3-5ppm, suitable for early leak identification in HVAC systems.
  • Easy to use: Simply expose the strip to the area you suspect has a leak; observe any color change for immediate results.
  • Designed for checking ammonia levels in various systems.
  • 100 strips that are sensitive to ammonia vapors, showing a color change to pink when ammonia is detected. Convenient for routine checks, helping to monitor ammonia levels in environments such as refrigeration and air conditioning systems, as well as industrial settings.
  • Simple and direct to use, these strips provide visual feedback that is quick and easy to interpret, aiding in the regular maintenance of systems that use ammonia.
  • This product is not intended to treat, diagnose, or prevent any diseases and should not be used for medical purposes. It is intended for monitoring and maintenance purposes only.

Specifications

Color White
pH Range 5.5 to 6.5
For Use With (Application) Ammonia Detection in Solution
For Use With (Equipment) Refrigerators, Air Conditioners, HVAC Maintenance
Quantity 100 Pk.
Range 3 to 5 ppm
Type Phenolphthalein Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.