Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein TS, Reagents

Catalog No. 01334110
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-110 500 mL
01-334-114 100 mL
01-334-112 250 mL
01-334-111 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-334-110 Supplier Reagents Holdings Llc Supplier No. CP260700500A
Only null left
Add to Cart
Add to Cart

Phenolphthalein TS, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 77°C
Grade USP
Packaging Poly Bottle
Specific Gravity 0.7861
Color Colorless
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Phenolphthalein TS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.