Learn More
Phenomenex Inc Phenomenex Lysis-Loading Buffer, Guanidine HCl 57%, Triton X-100, Urea
Supplier: Phenomenex Inc AL08509A
Clarity OTX Lysis-Loading Buffer is a guanidine hydrochloride-based lysis and loading reagent formulated for sample lysis and loading workflows. The formulation combines chaotropic agents, detergent, and reducing agents to denature biomolecules and assist sample preparation and loading.
- Form: Liquid mixture.
- Composition: guanidine hydrochloride 57.3%; water 27.87%; urea 12.02%; Triton X-100 2%; citric acid 0.42%; TCEP (hydrochloride) 0.29%; L-cysteine 0.10%.
- Color: Colorless.
- Odor: Characteristic.
- Intended use: Lysis-loading buffer for sample preparation.
- Storage: keep container tightly closed in a well-ventilated place; store in original container; store locked up.
- Safety: harmful if swallowed; causes skin irritation; causes serious eye irritation.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.