Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxybenzamine Hydrochloride, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18605107 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-107 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-107 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1579500GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

P1579, 63-92-3, C18H22CINO·HCl

Spectrum™ Chemical Phenoxybenzamine Hydrochloride, USP is an alpha blocker used to treat high blood pressure and heavy sweating due to a certain tumor of the adrenal glands. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 63-92-3
Molecular Formula C18H23Cl2NO
Molecular Weight (g/mol) 340.29
InChI Key VBCPVIWPDJVHAN-UHFFFAOYNA-N
IUPAC Name hydrogen benzyl(2-chloroethyl)(1-phenoxypropan-2-yl)amine chloride
SMILES [H+].[Cl-].CC(COC1=CC=CC=C1)N(CCCl)CC1=CC=CC=C1

Specifications

CAS Max % 1
Loss on Drying 0.005
Melting Range 136-141°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 101%
Grade USP
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.