Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 1-Hydroxy-2-naphthoate 98.0+%, TCI America™
SDP

Catalog No. H079725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H079725G 25 g
H0797500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H079725G Supplier TCI America Supplier No. H079725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-54-7
Molecular Formula C17H12O3
Molecular Weight (g/mol) 264.28
MDL Number MFCD00014310
InChI Key QHDYIMWKSCJTIM-UHFFFAOYSA-N
Synonym phenyl 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthoic acid phenyl ester, 2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester, phenyl 1-hydroxy-2-naphthalenecarboxylate, unii-0749hk9b6b, phenyl-1-hydroxy-2-naphthate, phenyl-1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid phenyl ester, aon phenyl ester, 1-hydroxy-2-naphthoic
PubChem CID 67236
IUPAC Name phenyl 1-hydroxynaphthalene-2-carboxylate
SMILES C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 25 g
Formula Weight 264.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 1-Hydroxy-2-naphthoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.