Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl 2,3,4-Tri-O-benzyl-1-thio-beta-L-fucopyranoside 98.0+%, TCI America™
SDP

Catalog No. P18421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18421G 1 g
P18425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18421G Supplier TCI America Supplier No. P18421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 167612-35-3
Molecular Formula C33H34O4S
Molecular Weight (g/mol) 526.691
InChI Key WGAOZACWMVPQOY-NBOLALRDSA-N
PubChem CID 44629997
IUPAC Name (2S,3R,4S,5S,6R)-2-methyl-3,4,5-tris(phenylmethoxy)-6-phenylsulfanyloxane
SMILES CC1C(C(C(C(O1)SC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Formula Weight 526.69
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2,3,4-Tri-O-benzyl-1-thio-beta-L-fucopyranoside
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.