Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Phenyl Salicylate
SCI_ED

Catalog No. S25469 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25469 100 g Solid
S25469A 500 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25469

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 118-55-8
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
InChI Key ZQBAKBUEJOMQEX-UHFFFAOYSA-N
Synonym phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate
PubChem CID 8361
ChEBI CHEBI:34918
IUPAC Name phenyl 2-hydroxybenzoate
SMILES C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Color White
Density 1.25g/mL
Assay Percent Range 99%
Packaging Solid
Linear Formula 2·HOC6H4·1·CO2C6H5
Quantity 100 g
Formula Weight 214.22
Chemical Name or Material Phenyl Salicylate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.