Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Phenyl Sepharose™ CL-4B Gel

Catalog No. 45000145 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-145 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-145 Supplier Cytiva Supplier No. 17081001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For immunoglobulin purification and fractionation as well as immunoprecipitation procedures

  • Strong binding to Fc region of IgG
  • Excellent for polyclonal antibody purification
  • High capacity: 20mg human IgG/mL

TRUSTED_SUSTAINABILITY

Specifications

Product Type Phenyl Sepharose CL-4 B
Flow Rate <150 cm/hr
For Use With (Application) For Hydrophobic Interaction Chromato Graphy
Quantity 200 mL
Ligand Type Phenyl
Particle Size 45 μm-165 μm
pH Range 3 to 12
Support Type 4% Cross-linked Agarose
Storage Requirements 4°C to 30°C, 20% Ethanol or 0.1 M NaOH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.