Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Trimethylsilylmethyl Sulfone 95.0+%, TCI America™
SDP

Catalog No. P12505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12505G 5 g
P12501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12505G Supplier TCI America Supplier No. P12505G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17872-92-3
Molecular Formula C10H16O2SSi
Molecular Weight (g/mol) 228.38
MDL Number MFCD00012364
InChI Key OEXJFEDAGJGOEP-UHFFFAOYSA-N
Synonym (Phenylsulfonylmethyl)trimethylsilane
PubChem CID 3290840
IUPAC Name [(benzenesulfonyl)methyl]trimethylsilane
SMILES C[Si](C)(C)CS(=O)(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 228.38
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Phenyl Trimethylsilylmethyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.