Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylacetaldehyde, 95%

Catalog No. AAA1426322
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1426322 100 g
AAA1426314 25 g
AAA1426336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1426322 Supplier Thermo Scientific Chemicals Supplier No. A1426322
Add to Cart

CAS: 122-78-1 | C8H8O | 120.15 g/mol

Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.

Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-78-1
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00006993
InChI Key DTUQWGWMVIHBKE-UHFFFAOYSA-N
Synonym phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde
PubChem CID 998
ChEBI CHEBI:16424
IUPAC Name 2-phenylacetaldehyde
SMILES O=CCC1=CC=CC=C1

Specifications

Density 1.075
Boiling Point 194°C to 196°C
Flash Point 68°C (154°F)
Refractive Index 1.535
Quantity 100 g
Beilstein 385791
Sensitivity Air sensitive
Merck Index 14,7265
Solubility Information Miscible with alcohol and diethyl ether. Slightly miscible with water.
Formula Weight 120.15
Percent Purity 95%
Chemical Name or Material Phenylacetaldehyde
Show More Show Less
Hazard Category H227-H302-H317
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c
EINECSNumber 204-574-5
RTECSNumber CY1420000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.