Learn More
Sigma Organic Chemistry Phenylacetamidoxime | 1G | MFCD00019953
Supplier: Sigma Organic Chemistry BOG001181G

Phenylacetamidoxime , 1G
About this Item:
MDL #: MFCD00019953
Molecular Weight: 150.18
UNSPSC Code: 12352200
Chemical Formula: C8H10N2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.