Learn More
Bioassay Systems EnzyChrom™ Phenylalanine Assay Kit. For the quantitative determination of L-phenylalanine and evaluation of drug effects on its metabolism.
SDP
Catalog No. NC1438107 Shop All Bioassay Systems Products
Encompass_Preferred
For the quantitative determination of L-phenylalanine and evaluation of drug effects on its metabolism. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. Linear detection range of 2 - 300 µM L-phenylalanine. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/595nm; Samples: Serum, urine etc; Size: 100 tests.
Catalog No. NC1438107 Supplier Bioassay Systems Supplier No. EPHE100
For the quantitative determination of L-phenylalanine and evaluation of drug effects on its metabolism. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. Linear detection range of 2 - 300 µM L-phenylalanine. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL530/595nm; Samples: Serum, urine etc; Size: 100 tests.