Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylethyl Alcohol, 98%, Spectrum™ Chemical
SDP

Catalog No. 18612302 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
15.9 kg
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-302 200 L Amber Glass Bottle
18-612-301 100 g Amber Glass Bottle
18-612-300 1 kg Amber Glass Bottle
18-612-303 15.9 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-302 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1057200LTBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 60-12-8

Specifications

CAS Max % 1
Boiling Point 218°C (424.4°F)
Packaging Amber Glass Bottle
Quantity 200 L
Specific Gravity 1.017 - 1.019
Molecular Weight (g/mol) 122.17
Percent Purity 0.98
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Phenylethyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.