Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylethyl caffeate, MP Biomedicals™

Catalog No. ICN19377510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19377510 10 mg
ICN19377580 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19377510 Supplier MP Biomedicals, Inc Supplier No. 0219377510

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Phenethyl caffeate is a cytotoxic agent to cancer cell lines. It acts as an inhibitor of protein tyrosine kinase and ornithine decarboxylase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104594-70-9
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.31
MDL Number MFCD00866470
InChI Key SWUARLUWKZWEBQ-VQHVLOKHSA-N
Synonym caffeic acid phenethyl ester, phenethyl caffeate, cape, capeee, phenethyl 3-3,4-dihydroxyphenyl acrylate, phenylethyl caffeate, unii-g960r9s5sk, caffeic acid phenylethyl ester, 2-phenylethyl caffeate, caffeic acid 2-phenylethyl ester
PubChem CID 5281787
ChEBI CHEBI:8062
IUPAC Name 2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O

Specifications

Color White
Quantity 10 mg
Formula Weight 284.3
Physical Form Powder
Chemical Name or Material Phenylethyl caffeate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.