CAS RN 104594-70-9
Phenylethyl 3,4-dihydroxycinnamate, 98+%
CAS: 104594-70-9 Molecular Formula: C17H16O4 Molecular Weight (g/mol): 284.31 MDL Number: MFCD00866470 InChI Key: SWUARLUWKZWEBQ-VQHVLOKHSA-N Synonym: caffeic acid phenethyl ester,phenethyl caffeate,cape,capeee,phenethyl 3-3,4-dihydroxyphenyl acrylate,phenylethyl caffeate,unii-g960r9s5sk,caffeic acid phenylethyl ester,2-phenylethyl caffeate,caffeic acid 2-phenylethyl ester PubChem CID: 5281787 ChEBI: CHEBI:8062 IUPAC Name: 2-phenylethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate SMILES: OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O
MilliporeSigma™ Calbiochem™ CAPE
A cell-permeable active component of propolis from honeybee hives.
Phenethyl Caffeate 98.0+%, TCI America™
CAS: 104594-70-9 Molecular Formula: C17H16O4 Molecular Weight (g/mol): 284.31 MDL Number: MFCD00866470 InChI Key: SWUARLUWKZWEBQ-VQHVLOKHSA-N Synonym: caffeic acid phenethyl ester,phenethyl caffeate,cape,capeee,phenethyl 3-3,4-dihydroxyphenyl acrylate,phenylethyl caffeate,unii-g960r9s5sk,caffeic acid phenylethyl ester,2-phenylethyl caffeate,caffeic acid 2-phenylethyl ester PubChem CID: 5281787 ChEBI: CHEBI:8062 IUPAC Name: 2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate SMILES: OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O
R&D Systems™ Caffeic acid phenethyl ester
CAS: 104594-70-9 Molecular Formula: C17H16O4
MP Biomedicals, Inc Phenylethyl caffeate, MP Biomedicals™
CAS: 104594-70-9 Molecular Formula: C17H16O4 Molecular Weight (g/mol): 284.31 MDL Number: MFCD00866470 InChI Key: SWUARLUWKZWEBQ-VQHVLOKHSA-N Synonym: caffeic acid phenethyl ester,phenethyl caffeate,cape,capeee,phenethyl 3-3,4-dihydroxyphenyl acrylate,phenylethyl caffeate,unii-g960r9s5sk,caffeic acid phenylethyl ester,2-phenylethyl caffeate,caffeic acid 2-phenylethyl ester PubChem CID: 5281787 ChEBI: CHEBI:8062 IUPAC Name: 2-phenylethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate SMILES: OC1=CC=C(\C=C\C(=O)OCCC2=CC=CC=C2)C=C1O