Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylmercury(II) chloride, 96%, Hg 63.5%

Catalog No. AA3711309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3711309 10 g
AA3711318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3711309 Supplier Thermo Scientific Chemicals Supplier No. 03711309
Add to Cart

CAS: 100-56-1 | C6H5ClHg | 313.15 g/mol

The reaction of phenylmercury (II) chloride with acetophenonethiosemicarbazone (Hatsc) in ethanol in 1: 1 mole ratio undergoes symmetrisation forming the products, HgCl2 (Hatse) 2 and Ph2Hg instead of the anticipated compound PhHgCl (Hatse). A new method for the determination of chloride ion is based on the formation of phenylmercury(II) chloride, its extraction into chloroform and reaction with sodium diethyldithiocarbamate to form phenylmercury(II) diethyldithiocarbamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-56-1
Molecular Formula C6H5ClHg
Molecular Weight (g/mol) 313.15
MDL Number MFCD00000463
InChI Key AWGTVRDHKJQFAX-UHFFFAOYSA-M
Synonym phenylmercuric chloride, agrenal, phenylmercury chloride, mercury, chlorophenyl, hexason, mercutal, merfazin, stopspot, agronal, mersolite 2
PubChem CID 7511
IUPAC Name chloro(phenyl)mercury
SMILES Cl[Hg]C1=CC=CC=C1

Specifications

Melting Point 246°C to 250°C
Color White
Quantity 10 g
UN Number UN2026
Merck Index 14,7301
Solubility Information Soluble in benzene, ether, pyridine; Very slightly soluble in water
Formula Weight 313.18
Percent Purity 96%
Concentration or Composition (by Analyte or Components) Hg 63.5%
Physical Form Powder
Chemical Name or Material Phenylmercury(II) chloride
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLMERCURIC COMPOUNDS, N.O.S.
EINECSNumber 202-865-1
RTECSNumber OW1400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.